Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93203

Supelco

Carbamazepine 10,11-epoxide

analytical standard

Synonym(s):

1a,10b-Dihydro-6H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

NC(=O)N1c2ccccc2C3OC3c4ccccc14

InChI

1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)

InChI key

ZRWWEEVEIOGMMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mona Darwish et al.
Clinical therapeutics, 37(2), 325-337 (2014-12-03)
Polypharmacy is common in psychiatry practice and can lead to an increased risk of drug interactions. Armodafinil, a wakefulness-promoting agent, has been studied as adjunctive therapy for the treatment of major depressive episodes associated with bipolar I disorder. Armodafinil and
Ramesh Boinpally et al.
Clinical therapeutics, 36(11), 1638-1649 (2014-09-23)
Vilazodone is a serotonin reuptake inhibitor and 5-HT1A partial agonist approved for the treatment of major depressive disorder in adults. Vilazodone seems to be metabolized primarily by the cytochrome P-450 (CYP) 3A4 isozyme and non-CYP-mediated pathways; concomitant use of drugs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service