Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900640

Sigma-Aldrich

1,4-Dioxane

anhydrous, ZerO2®, 99.8%

Synonym(s):

1,4-Dioxane ZerO2®, Degassed and low oxygen 1,4-dioxane, Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.21
grade:
ZerO2®
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

ZerO2®

Quality Level

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

99.8%

form

liquid

autoignition temp.

356 °F

quality

anhydrous

expl. lim.

22 %

impurities

≤0.003% water
water

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service