Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

1.09671

Supelco

1,4-Dioxane

for analysis EMSURE® ACS,ISO

Synonym(s):

1,4-Dioxane, Glycolethylether, 1,4-Diethylene dioxide, 1,4-Dioxacyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8
NACRES:
NB.21
grade:
ACS reagent
for analytical purposes
bp:
101.5 °C/1013 hPa
vapor pressure:
41 hPa ( 20 °C)

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. ISO

vapor pressure

41 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

300 °C

potency

5200 mg/kg LD50, oral (Rat)
7600 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-25.2 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0008 meq/g Acidity
≤0.005% Acetaldehyde
≤0.005% Peroxide (as H2O2)
≤0.01% Carbonyl compounds (as CO)
≤0.05% Formaldehyde
≤0.05% Water

evapn. residue

≤0.001%

color

APHA: ≤10

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

O1CCOCC1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Preferential Solvation Phenomena as Solute-Induced Structure-Making/Breaking Processes: Linking Thermodynamic Preferential Interaction Parameters to Fundamental Structure Making/Breaking Functions.: This study explores the solvation dynamics and interactions in solutions containing 1,4-dioxane, highlighting the solute′s influence on molecular arrangements and interactions (Chialvo AA, 2024).

  • Green Synthesis of Highly Fluorescent NCQDs: A Comprehensive Study on Synthesis, Characterization, Photophysical Properties, pH Sensing, Heavy Metal Detection, and Solvatochromic Behavior through Hydrothermal Method.: Discusses the development of nanocrystals using 1,4-dioxane as a solvent, focusing on their application in environmental monitoring and chemical sensing (Negi P et al., 2024).

  • Concentration Fluctuation/Microheterogeneity Duality Illustrated with Aqueous 1,4-Dioxane Mixtures.: This paper presents a theoretical analysis of the mixture behavior of water and 1,4-dioxane, providing insights into the molecular interactions and solution properties (Kolaříková A et al., 2024).

  • Synthesis, molecular docking studies and biological evaluation of N-(4-oxo-2-(trifluoromethyl)-4H-chromen-7-yl) benzamides as potential antioxidant, and anticancer agents.: Explores the synthesis of new compounds using 1,4-dioxane and their potential as therapeutic agents, highlighting the versatility of 1,4-dioxane in drug development (Jorepalli S et al., 2024).


Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service