Skip to Content
MilliporeSigma
1,4-Dioxane SAJ first grade

1,4-Dioxane

1,4-Dioxane SAJ first grade
Synonym(s):
Diethylene oxide, Dioxane, 1,4-Dioxane
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
Technique