Skip to Content
MilliporeSigma
All Photos(1)

Documents

85927

Millipore

ECC ChromoSelect Selective Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

E. coli-Coliform Selective Agar ChromoSelect, ECC Selective Agar ChromoSelect

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 10.0 g/L
casein enzymic hydrolysate, 3.3 g/L
chromogenic mixture, 0.43 g/L
disodium hydrogen phosphate, 1 g/L
peptone, special, 6.0 g/L
sodium chloride, 2 g/L
sodium dihydrogen phosphate, 0.6 g/L
sodium pyruvate, 1 g/L
sorbitol, 1 g/L
Tergitol
7, 0.15 g/L

tryptophan, 1 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Citrobacter spp.
selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Looking for similar products? Visit Product Comparison Guide

Application

ECC ChromoSelect Selective Agar is a selective medium recommended for the simultaneous detection of Escherichia coli and coliforms in water and food samples. The ingredients help even the sublethally injured coliforms to grow rapidly, Tergitol inhibits Gram-positive as well as some Gram-negative bacteria other than coliforms. The chromogenic mixture contains two chromogenic substrates as Salmon-GAL and X-glucuronide. The enzyme β-D-galactosidase produced by coliforms cleaves Salmon-GAL, resulting in the salmon-to-red coloration of coliform colonies. The enzyme β-D-glucuronidase produced by E. coli, cleaves X-glucuronide. E. coli forms dark blue-to-violet colored colonies due to cleavage of both Salmon-GAL and X-glucuronide. The addition of tryptophan improves the indole reaction. To confirm E. coli, add a drop of Kovac′s reagent (Catalog No. 60983) on the dark blue-to-violet colony. Formation of cherry-red color indicates the positive reaction.

Preparation Note

Suspend 26.5 g in 1 litre distilled water. Heat in a boiling water bath or in a free flowing steam, with stirring to dissolve the medium completely (approximately 35 minutes). Medium may show haziness, but it does not affect the performance. If desired to inhibit Pseudomonas and Aeromonas species add 5 mg Cefsulodin (Cat. No. 22126) for surface and pour plate method and 10 mg Cefsulodin for membrane filter technique. Mix well.

Reconstitution

Suspend 26.5 g in 1 liter of distilled water. Heat in a boiling water bath or in a free flowing steam, with stirring to dissolve the medium completely (approximately 35 minutes). Medium may show haziness, but it does not affect the performance. If desired to inhibit Pseudomonas and Aeromonas species, add 5 mg of cefsulodin for surface and pour plate method and 10 mg of cefsulodin for membrane filter technique. Mix well.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongda Wang et al.
Light, science & applications, 9, 118-118 (2020-07-21)
Early identification of pathogenic bacteria in food, water, and bodily fluids is very important and yet challenging, owing to sample complexities and large sample volumes that need to be rapidly screened. Existing screening methods based on plate counting or molecular

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service