Skip to Content
MilliporeSigma

09142

ECD ChromoSelect Agar with MUG

NutriSelect® Plus, suitable for microbiology

Synonym(s):

ECD Agar ChromoSelect with MUG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L , chromogenic substrate, 0.10 g/L , dipotassium hydrogen phosphate, 4.0 g/L , fluorogenic substrate, 0.07 g/L , lactose, 5.0 g/L , potassium dihydrogen phosphate, 1.5 g/L , sodium chloride, 5.0 g/L , synthetic detergent (bile salt replacement), 1.5 g/L , tryptone (vegetable), 20.0 g/L , tryptophan, 1.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
water monitoring, microbiology

suitability

selective and differential for Enterococcus spp., selective and differential for Escherichia coli, selective and differential for Klebsiella spp., selective and differential for coliforms

Application

For the detection of Escherichia coli in water and food samples by using a combination of chromogenic and fluorogenic substrate.

Preparation Note

Suspend 53.17 g in 1000 ml distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library