Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78698

Supelco

Sodium bicarbonate/Sodium carbonate regenerant

Na2CO3 70 mM and NaHCO3 70 mM in water, IC regenerant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

Quality Level

form

liquid

quality

Na2CO3 70 mM and NaHCO3 70 mM in water

shelf life

limited shelf life, expiry date on the label

concentration

70 mM Na2CO3 in H2O
70 mM NaHCO3 in H2O

technique(s)

ion chromatography: suitable

Application

Metrohm IC application note AN-CS-003: Biogenic amines in addition to standard cations with conductivity detection following sequential suppression and Dose-in Gradient.

Metrohm IC application note AN-CS-010: Traces of lithium and sodium besides monoethanolamine in water-steam circuits of thermal power plants.

Metrohm IC application note AN-CS-006: Aliphatic amines and ethanol amines in addition to standard cations by sequential suppression.

Linkage

to be used in combination with eluent concentrate for Metrosep C Supp 1 and 2: 78737

Visit the IC Portal to learn more

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service