Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43617

Supelco

Sodium hydroxide solution

0.1 M NaOH in water (0.1N), Eluent concentrate for IC

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NB.21

Product Name

Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC

Agency

suitable for SM 4500 - NH3

Quality Level

vapor pressure

3 mmHg ( 37 °C)

form

liquid

concentration

0.1 M NaOH in water (0.1N)

technique(s)

ion chromatography: suitable

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-S-294: Phosphor species in whitening toothpaste.
Sodium hydroxide concentrate was used in deposition-precipitation method to adjust pH during synthesis of platinum systems to test for liquid-phase selective hydrogenation of crotonaldehyde to crotyl alcohol.

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with sodium hydroxide and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesús Hidalgo-Carrillo et al.
Journal of colloid and interface science, 382(1), 67-73 (2012-07-06)
Natural phosphates are an important natural resource in Morocco, which needs to be valorised. They can be used not only as fertilisers but also as catalysts. Here, we report for the first time on its use as support for platinum
Nicole von Burg et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(35), 12835-12840 (2014-08-20)
Group 3 innate lymphoid cells (ILC3s) have emerged as important cellular players in tissue repair and innate immunity. Whether these cells meaningfully regulate adaptive immune responses upon activation has yet to be explored. Here we show that upon IL-1β stimulation
Richard J Travers et al.
Blood, 124(22), 3183-3190 (2014-09-10)
Polyphosphate (polyP) is secreted by activated platelets and has been shown to contribute to thrombosis, suggesting that it could be a novel antithrombotic target. Previously reported polyP inhibitors based on polycationic substances, such as polyethylenimine, polyamidoamine dendrimers, and polymyxin B
Horacio Cardenas et al.
Epigenetics, 9(11), 1461-1472 (2014-12-04)
A key step in the process of metastasis is the epithelial-to-mesenchymal transition (EMT). We hypothesized that epigenetic mechanisms play a key role in EMT and to test this hypothesis we analyzed global and gene-specific changes in DNA methylation during TGF-β-induced
Valdeir Arantes et al.
Biotechnology for biofuels, 7, 87-87 (2014-07-01)
There is still considerable debate regarding the actual mechanism by which a "cellulase mixture" deconstructs cellulosic materials, with accessibility to the substrate at the microscopic level being one of the major restrictions that limits fast, complete cellulose hydrolysis. In the

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service