Skip to Content
MilliporeSigma

70145

Malt Extract Agar

NutriSelect® Plus, powder, for yeasts, for molds, pack of 100 g, pkg of 500 g, pkg of 2.5 kg, suitable for microbiology

Synonym(s):

Malt-based culture medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171605
NACRES:
NA.74
eCl@ss:
42040102
Manufacturer/tradename:
NutriSelect® Plus
Form:
powder
Suitability:
molds, selective by low pH for Aspergillus spp., selective by low pH for Candida spp., selective by low pH for Penicillium spp., selective by low pH for Pichia spp., selective by low pH for Saccharomyces spp., selective by low pH for Zygosaccharomyces spp., yeasts
Application(s):
agriculture
bioburden testing
clinical testing
environmental
food and beverages, microbiology
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L , malt extract, 30 g/L , mycological peptone, 5 g/L

packaging

pack of 100 g, pkg of 2.5 kg, pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages, microbiology

suitability

molds, selective by low pH for Aspergillus spp., selective by low pH for Candida spp., selective by low pH for Penicillium spp., selective by low pH for Pichia spp., selective by low pH for Saccharomyces spp., selective by low pH for Zygosaccharomyces spp., yeasts

General description

Malt extract agar is considered as a general growth media due to their richness and nutrient balance. It is suitable for the cultivation of fastidious microorganisms. With acidic pH, they are used for the isolation, cultivation and maintenance of yeast and molds. Malt extract contains polysaccharides which are used as energy source. Mycological peptone serves as a nitrogen source and agar is the solidifying agent rapidly yielding a luxuriant growth with typical morphology and pigmentation. Lactic acid, tartaric acid or antibiotics can be added to suppress bacterial growth and make the medium more selective. Reiss recommends a modified malt extract medium for the selective cultivation of Aspergillus flavus. According to Rapp, addition of certain indicator dyes to malt extract agar allows differentiation of yeast and bacterial colonies.

Application

Malt Extract Agar is recommended for the detection, isolation and enumeration of fungi, particularly yeasts and molds, in various materials and for the cultivation of the strains for microbiological vitamin assays

Preparation Note

Suspend 50 g in 1 litre of distilled water and bring to the boil to dissolve. Sterilize by autoclaving at 115°C for 10 minutes. For inhibition of bacterial growth, 2 ml of a 10% sterile filtered lactic acid can be added to adjust the pH to 3.5.

Other Notes

Biological attributes of colony-type variants of Candida albicans; Volatile metabolites and other indicators of P. aurantiogriseum - growth on different substrates
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Malt Extract Agar


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library