Skip to Content
MilliporeSigma
MES monohydrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

MES monohydrate

MES monohydrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid monohydrate
Linear Formula:
C6H13NO4S · H2O
CAS Number:
Molecular Weight:
213.25

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Technique