Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66807

Sigma-Aldrich

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride

suitable for fluorescence, BioReagent, ≥97.0% (HPLC)

Synonym(s):

1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, 1,3-Benzodioxole-5,6-diamine dihydrochloride, 5,6-Diamino-1,3-benzodioxole dihydrochloride, MDB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2 · 2HCl
CAS Number:
Molecular Weight:
225.07
Beilstein:
5641607
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥97.0% (HPLC)

form

solid

fluorescence

λex 367 nm; λem 445 nm in 0.1 M phosphate pH 7.0 (after derivatization with pyruvate)

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].Nc1cc2OCOc2cc1N

InChI

1S/C7H8N2O2.2ClH/c8-4-1-6-7(2-5(4)9)11-3-10-6;;/h1-2H,3,8-9H2;2*1H

InChI key

YXEJRYIEFFUUID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride is a fluorescence tag for ⍺-keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.

Application

Fluorogenic reagent for α-ketoacids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

may discolor on storage

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hara et al.
Analytical biochemistry, 164(1), 138-145 (1987-07-01)
A simple, rapid, and highly sensitive high-performance liquid chromatographic method is described for the determination of N-acetyl- and N-glycolylneuraminic acids in human and animal sera, glycoproteins, and glycolipids. The neuraminic acids, released by acid hydrolysis of these biological samples, are
Capillary Electrophoresis of Carbohydrates: From Monosaccharides to Complex Polysaccharides, 249-249 (2010)
Xiaoxiao Guo et al.
The Analyst, 145(13), 4512-4521 (2020-05-16)
The polysialyltransferases (polySTs) catalyse the polymerisation of polysialic acid, which plays an important role in tumour metastasis. While assays are available to assess polyST enzyme activity, there is no methodology available specifically optimised for identification and quantitative evaluation of potential
K Hamahashi et al.
Journal of tissue engineering and regenerative medicine, 9(1), 24-30 (2012-11-21)
The authors aimed to repair and regenerate articular cartilage with layered chondrocyte sheets, produced using temperature-responsive culture dishes. The purpose of this study was to investigate the humoral factors produced by layered chondrocyte sheets. Articular chondrocytes and synovial cells were
Antonietta Manna et al.
Brain and behavior, 5(7), e00334-e00334 (2015-07-30)
Deep brain gray matter (GM) structures are involved in several neurodegenerative disorders and are affected by aging. In this study, we investigated the potential relationship between levels of brain-derived neurotrophic factor (BDNF), a putative biomarker of age- and clinically relevant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service