Skip to Content
MilliporeSigma
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride suitable for fluorescence, BioReagent, ≥97.0% (HPLC)

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride suitable for fluorescence, BioReagent, ≥97.0% (HPLC)
Synonym(s):
1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, 1,3-Benzodioxole-5,6-diamine dihydrochloride, 5,6-Diamino-1,3-benzodioxole dihydrochloride, MDB
Empirical Formula (Hill Notation):
C7H8N2O2 · 2HCl
CAS Number:
Molecular Weight:
225.07

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application