Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900438

Sigma-Aldrich

Super yellow light-emitting PPV copolymer

Synonym(s):

Livilux®, PDY-132

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

Quality Level

mol wt

average Mn >400,000
average Mw >1,300,000

General description

Super yellow light emitting PPV copolymer can form a thin film of poly(1,4-phenylenevinylene) (PPV) with high efficiencies. It has conjugated polymers as the backbone that can be soluble in water upon the incorporation of lipophilic solubilizing linkers.

Application

Features:
  • soluble super yellow light-emitting PPV copolymer
  • excellent reproducibility
  • broad emission spectrum
  • very high efficiency
  • very low operation voltage/for PM 100000 cd/m2 pulsed <10 V
  • very long DC & AC lifetime

Typical performance measured in standard device stack (ITO/PEDOT: PSS 20 nm/PDY-132 80 nm/Ba 6 nm/Ag 150 nm):
  • max. efficiency: 11 Cd/A
  • V_on: 2.2 V
  • V @ 100 cd/m2: 3.0 V
  • V @ 1000 cd/m2: 4.3 V
  • max. power eff.: 10 lm/W
  • external quantum eff.: 5.3%
  • CIE1931 @ 100 cd/m2: (0.50, 0.49)
  • DC lifetime @ 100 cd/m2 @ RT: >>220000
  • AC lifetime @ 200 cd/m2 @ RT: 11000
PDY-132 can be used in the preparation of electroluminescent line material, which can be used for making line art by direct-write patterning. It can also be used as a fluorescent material with the absorption peak at 450 nm, that can be developed by blade coating on the substrates of the transfer printed micro-sized light emitting diodes (μLED).

Legal Information

Livilux is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Light-emitting polymers-The shape of things to come?
Burn PL and Samuel ID
Materials Today, 1(4), 3-5 (1998)
Transfer-printed micro-LED and polymer-based transceiver for visible light communications
Rae K, et al.
Optics Express, 26(24), 31474-31483 (2018)
Luminescent line art by direct-write patterning
Lindh EM, et al.
Light: Science & Applications, 5(3), e16050-e16050 (2016)

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service