Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

63086

Supelco

Magnesium ionophore III

Selectophore, function tested

Synonym(s):

ETH 4030, N,N′′-Octamethylene-bis(N′-heptyl-N′-methylmalonamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H58N4O4
CAS Number:
Molecular Weight:
538.81
Beilstein:
6016307
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

product line

Selectophore

quality

function tested

SMILES string

CCCCCCCN(C)C(=O)CC(=O)NCCCCCCCCNC(=O)CC(=O)N(C)CCCCCCC

InChI

1S/C30H58N4O4/c1-5-7-9-15-19-23-33(3)29(37)25-27(35)31-21-17-13-11-12-14-18-22-32-28(36)26-30(38)34(4)24-20-16-10-8-6-2/h5-26H2,1-4H3,(H,31,35)(H,32,36)

InChI key

MXMJPDBVBJZBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore used in solvent polymeric membrane electrodes for the assay of Mg2+ activities in blood serum

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Muller et al.
Mikrochimica Acta, III, 283-283 (1988)
M. Mai-Zurawska et al.
Analytica Chimica Acta, 218, 47-47 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service