Skip to Content
MilliporeSigma
Magnesium ionophore III Selectophore™, function tested

Magnesium ionophore III

Magnesium ionophore III Selectophore™, function tested
Synonym(s):
ETH 4030, N,N′′-Octamethylene-bis(N′-heptyl-N′-methylmalonamide)
Empirical Formula (Hill Notation):
C30H58N4O4
CAS Number:
Molecular Weight:
538.81

Products

Brand
Formula Weight
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing