Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63098

Supelco

Magnesium ionophore III - membrane A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

storage temp.

2-8°C

General description

Visit our Sensor Applications portal to learn more.

Packaging

box contains 5 membranes

Other Notes

Ion selective polymer membrane for assaying magnesium in blood, water etc

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Mai-Zurawska et al.
Analytica Chimica Acta, 218, 47-47 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service