Skip to Content
MilliporeSigma
All Photos(1)

Documents

60215

Supelco

Potassium citrate monobasic

≥98.0% (NT)

Synonym(s):

Citric acid monopotassium salt, Potassium dihydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2C6H5O7
CAS Number:
Molecular Weight:
230.21
Beilstein:
4772973
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.0% (NT)

form

solid

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O

InChI

1S/C6H8O7.K/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+1/p-1

InChI key

WKZJASQVARUVAW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna-Cathrine Neumann et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(2), 207-214 (2018-10-16)
Eutrophication of water bodies can promote cyanobacterial (blue-green algae) blooms, which has become a source of increasing concern for both recreational and drinking water use. Many bacterial species can produce toxins that pose threats to wildlife, domestic animals and humans.
Monica Viladomiu et al.
Cell host & microbe, 29(4), 607-619 (2021-02-05)
Adherent-invasive E. coli (AIEC) are enriched in the intestinal microbiota of patients with Crohn's disease (CD) and promote intestinal inflammation. Yet, how AIEC metabolism of nutrients impacts intestinal homeostasis is poorly defined. Here, we show that AIEC encoding the large subunit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service