Skip to Content
MilliporeSigma

Skip To

ED3SS

Ethylenediaminetetraacetic acid trisodium salt hydrate

≥95%

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid trisodium salt hydrate, EDTA trisodium salt hydrate, Edathamil hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g

Available to ship TODAYfromMILWAUKEE

$103.00
500 g
Check Cart for Availability
$297.00
1 kg

Available to ship TODAYfromMILWAUKEE

$512.00

About This Item

Empirical Formula (Hill Notation):
C10H13N2Na3O8 · xH2O
CAS Number:
Molecular Weight:
358.19 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
205-758-8
MDL number:

$103.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥95%

form

powder

reaction suitability

reagent type: chelator

mp

300 °C

solubility

1 M NaOH: soluble, clear, colorless to light yellow

storage temp.

room temp

SMILES string

O.[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.3Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;3*+1;/p-3

InChI key

KKTNQJMZBPLVKM-UHFFFAOYSA-K

Application

Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.

Other Notes

This is a physical blend of the disodium and tetrasodium salts of EDTA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E5391ED2PE4884
assay

≥95%

assay

≥99.0%

assay

≥98%

assay

99.0-101.0%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

solid

mp

300 °C

mp

-

mp

255-280 °C (dec.) (lit.)

mp

248 °C (dec.) (lit.)

solubility

1 M NaOH: soluble, clear, colorless to light yellow

solubility

H2O: 0.1 M, clear, colorless

solubility

3 M KOH: 160 mg/mL, clear to very slightly hazy, colorless to faintly yellow

solubility

H2O: 100 g/L at 20 °C

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters