Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53900

Sigma-Aldrich

Hydrazine sulfate salt

puriss. p.a., ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2SO4
CAS Number:
Molecular Weight:
130.12
Beilstein:
8778859
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0%

form

powder or crystals

ign. residue

≤0.05% (as SO4)

pH

1.3 (52 g/L)

mp

254 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤400 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NN.OS(O)(=O)=O

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)

InChI key

ZGCHATBSUIJLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hydrazine sulfate salt can be used as a reactant to synthesize sulfonated polyoxadiazoles in the presence of non-sulfonated diacids in polyphosphoric acid. It is also used as a modifier in the functionalization of microcrystalline cellulose, which is used as an adsorbent for the removal of dyes from water.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold nanoparticles formation in the aqueous system of gold (III) chloride complex ions and hydrazine sulfate-kinetic studies.
Streszewski B, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 397, 63-72 (2012)
Proton magnetic resonance spectra of hydrazine salts. Part 1.
Pratt L and Richards RE.
Transactions of the Faraday Society, 49, 744-751 (1953)
Tumorigenesis studies with 1,2-dimethylhydrazine dihydrochloride, hydrazine sulfate, and isonicotinic acid in golden hamsters.
B Toth
Cancer research, 32(4), 804-807 (1972-04-01)
Azo Compounds. 1 A Re-examination of the Structure of the Product from the Reaction of Heptane-2, 6-dione with Hydrazine Sulfate and Sodium Cyanide in Dilute Solution.
Overberger CG and Marks BS.
Journal of the American Chemical Society, 77(15), 4097-4100 (1955)
Boran Kartal et al.
FEMS microbiology reviews, 37(3), 428-461 (2012-12-06)
Anaerobic ammonium-oxidizing (anammox) bacteria primarily grow by the oxidation of ammonium coupled to nitrite reduction, using CO2 as the sole carbon source. Although they were neglected for a long time, anammox bacteria are encountered in an enormous species (micro)diversity in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service