Skip to Content
MilliporeSigma
All Photos(5)

Documents

207942

Sigma-Aldrich

Hydrazine monohydrate

N2H4 64-65 %, reagent grade, ≥97%

Synonym(s):

Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2O
CAS Number:
Molecular Weight:
50.06
Beilstein:
878137
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 25 °C)

Assay

≥97%

form

liquid

composition

N2H4, 64-65%

expl. lim.

99.99 %

refractive index

n20/D 1.428 (lit.)

pH

10.6-10.7 (10 g/L)

bp

120.1 °C (lit.)

mp

−51.7 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

O.NN

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of hydrazine monohydrate exhibit triclinic crystal system and space group P3121.

Application

Hydrazine monohydrate has been used to synthesize 2-(6-ethylamino-3-ethylimino-2,7-dimethyl-3H-xanthen-9-yl)benzoic acid hydrazide and vinyl-benzylamine hydrochloride. It may also be used as a source of hydrogen via decomposition in the presence of supported bimetallic catalyst composed of Ni-Pt alloy nanoparticles and mesoporous CeO2 support.
Hydrazine monohydrate may be used as a reducing agent for the reduction of:
  • Nitroarenes in the presence of hydrogen peroxide functionalized carbon nanotubes.
  • 4-Iodo-3-nitroanisole to form 2-iodo-5-methoxyaniline.
  • Graphene oxide (GO) to form graphene.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup - (anhydrous substance)

Flash Point(C)

38 °C - closed cup - (anhydrous substance)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convergent synthesis of the quinolone substructure of BILN 2061 via carbonylative Sonogashira coupling/cyclization.
Haddad N, et al.
The Journal of Organic Chemistry, 71(13), 5031-5034 (2006)
Yuan-Yuan Jiang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(43), 15439-15445 (2015-10-17)
The development of active, selective, and robust catalysts is a key issue in promoting the practical application of hydrazine monohydrate (N2 H4 ⋅H2 O) as a viable hydrogen carrier. Herein, the synthesis of a supported Ni-Pt bimetallic nanocatalyst on mesoporous
High performance supercapacitor for efficient energy storage under extreme environmental temperatures.
Vellacheri R, et al.
Nano Energy, 8, 231-237 (2014)
Carbon nanotubes oxidized by a green method as efficient metal-free catalysts for nitroarene reduction.
Wu S, et al.
Physical Chemistry Chemical Physics, 17(3), 1567-1571 (2015)
The crystal structure of hydrazine monohydrate.
Liminga R and Olovsson I.
Acta Crystallographica, 17(12), 1523-1528 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service