Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04603

Supelco

Hydrazinium sulfate

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Hydrazinium sulfate, Hydrazonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H6N2O4S
CAS Number:
Molecular Weight:
130.12
MDL number:
UNSPSC Code:
12352301
EC Index Number:
233-110-4
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

potency

601 mg/kg LD50, oral (Rat)

pH

1.5 (20 °C, 50 g/L in H2O)

mp

254 °C (decomposition)

density

1.37 g/cm3 at 20 °C

bulk density

450 kg/m3

storage temp.

no temp limit

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)

InChI key

ZGCHATBSUIJLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service