Skip to Content
MilliporeSigma

Skip To

2105

Sodium hydroxide solution

0.1 N

Synonym(s):

Caustic Soda Solution, NaOH Solution, Sodium Hydroxide Liquid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352316
NACRES:
NA.26
PubChem Substance ID:
Concentration:
0.1 N
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

0.1 N

impurities

endotoxin, free

General description

Sodium hydroxide is a deliquescent, white crystalline solid. It is an extensively used industrial alkali. It is used for adjusting the pH values of water during instrumentation and characterization.

Application

Sodium hydroxide solution is suitable for use with E-TOXATE Kits, ET0100, ET0200, and ET0300 as pH adjustment solution. Avoid contamination.

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
72064415413S2770
description

0.1 N

description

49-51% in water, eluent for IC

description

50% in H2O

description

1.0 N, BioReagent, suitable for cell culture

concentration

0.1 N

concentration

49-51% in water

concentration

48-52% (by HCl, titration), 50% in H2O

concentration

1.0 N

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

form

liquid

form

liquid

form

liquid (viscous)

form

solution

impurities

endotoxin, free

impurities

≤0.01% ammonium hydroxide precipitate, ≤0.0005% total nitrogen (N)

impurities

-

impurities

-


Still not finding the right product?

Explore all of our products under Sodium hydroxide solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library