Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422096

Supelco

AmberChrom 50WX4 Ion Exchange Resin

hydrogen form, 100-200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

AmberChrom 50WX4 hydrogen form, hydrogen form, 100-200 mesh

description

cross-linkage 4%

Quality Level

form

beads

moisture

64-72%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

100-200 mesh

capacity

1.1 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dowex 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex 50WX4 hydrogen form can be used as a column packing material which is used in anion exchange chromatography.

Application

Dowex 50WX4 hydrogen form was used:
  • to investigate the effect of particle size on drug loading and release profiles
  • for the conversion of D,L-Glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate
Strongly acidic cation exchange resin that meets requirements of FDA Food Additive Regulation 21 CFR 173.25
Used for the recovery of useful radioactive species from waste nuclear material.

Other Notes

This strongly acidic cation exchange resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohapatra, P.K., et al.
Solvent Extr. Ion Exch., 22, 267-267 (2004)
Superacid polymers: Synthesis and analysis of AlCl3-sulfonic acid resin complexes.
Magnotta, V. L., and B. C. Gates.
Journal of Polymer Science Part A: Polymer Chemistry, 15, 1341-1347 (1977)
Maybelle K Go et al.
Biochemistry, 48(24), 5769-5778 (2009-05-12)
Product distributions for the reaction of glycolaldehyde labeled with carbon-13 at the carbonyl carbon ([1-(13)C]-GA) catalyzed by triosephosphate isomerase (TIM) in D(2)O at pD 7.0 in the presence of phosphite dianion and in its absence were determined by (1)H NMR
Sebastian Reichau et al.
The Journal of biological chemistry, 286(18), 16197-16207 (2011-04-02)
Tuberculosis remains a serious global health threat, with the emergence of multidrug-resistant strains highlighting the urgent need for novel antituberculosis drugs. The enzyme 3-deoxy-D-arabino-heptulosonate 7-phosphate synthase (DAH7PS) catalyzes the first step of the shikimate pathway for the biosynthesis of aromatic
Seong Hoon Jeong et al.
International journal of pharmaceutics, 361(1-2), 26-32 (2008-06-21)
Complexes of ion-exchange resins and dextromethorphan (DM), a model drug, were prepared using different methods including a batch and a column method with different functional groups, ion-exchange capacity, degree of crosslinking, and resin particle size. Drug loading efficiency, release profiles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service