Skip to Content
MilliporeSigma

217425

AmberChrom 1X8 Ion Exchange Resin

chloride form, 100-200 mesh

Synonym(s):

Dowex 1X8-200 Ion Exchange Resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 1X8 chloride form, chloride form, 100-200 mesh

Quality Segment

description

cross-linkage 8%

form

beads

crosslinking

8 % cross-linked

parameter

66 °C OH- form max. temp.

moisture

39-45%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

100-200 mesh

operating pH

0-14

capacity

1.2 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

General description

Dowex 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation.

Application

Dowex 1X8 chloride form was used for separation procedure by Dowex chromatography.
Strongly basic (type I) anion exchange resin for fine chemical processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

strongly basic anion.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.


Still not finding the right product?

Explore all of our products under AmberChrom 1X8 chloride form


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library