Skip to Content
MilliporeSigma

217514

AmberChrom™ 50WX8 Ion Exchange Resin

hydrogen form, 200-400 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amberchrom™ 50WX8, 200-400 mesh, hydrogen form

description

cross-linkage 8%

Quality Segment

form

beads

moisture

50-58%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

200-400 mesh

capacity

1.7 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

Amberlite™ HPR4800 Cl hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted. Amberlite™ HPR4800 Cl hydrogen form is used as catalyst. It is used in deionizing supernatant fraction samples for paper chromatography.

Application

Amberlite™ HPR4800 Cl hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly acidic cation exchange resin for fine chemical and pharmaceutical separations; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

strongly acidic cation.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.
This resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.
Amberlite is a trademark of DuPont de Nemours, Inc.


Still not finding the right product?

Explore all of our products under Amberchrom™ 50WX8


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library