Skip to Content
MilliporeSigma
Potassium antimonyl tartrate trihydrate purum p.a., 99.0-103% (RT)

Potassium antimonyl tartrate trihydrate

Potassium antimonyl tartrate trihydrate purum p.a., 99.0-103% (RT)
Synonym(s):
Antimony potassium tartrate trihydrate, Tartar emetic
Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · 3H2O
CAS Number:
Molecular Weight:
667.87
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade