Skip to Content
MilliporeSigma
All Photos(2)

Documents

36181

Supelco

Omethoate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12NO4PS
CAS Number:
Molecular Weight:
213.19
Beilstein:
1785256
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CNC(=O)CSP(=O)(OC)OC

InChI

1S/C5H12NO4PS/c1-6-5(7)4-12-11(8,9-2)10-3/h4H2,1-3H3,(H,6,7)

InChI key

PZXOQEXFMJCDPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yanhong Bai et al.
Journal of agricultural and food chemistry, 57(14), 6238-6245 (2009-06-19)
A practical, inexpensive, and green chemical process is greatly needed for degrading pesticides in food and environmental water. In this work, the impact of O(2) plasma treatment on reduction of dichlorvos (DDVP) and omethoate in maize was determined by gas
Ozden Tacal et al.
Journal of applied toxicology : JAT, 30(5), 469-475 (2010-03-17)
Organophosphorus pesticides used most commonly in Turkey include methamidophos, dichlorvos, O-methoate and diazinon. These toxic chemicals or their metabolites make a covalent bond with the active site serine of butyrylcholinesterase. Our goal was to identify the adducts that result from
Musa Isnas et al.
Toxicology and industrial health, 28(1), 27-34 (2011-10-26)
The study aimed mainly to assess effects of omethoate on certain oxidative stress biomarkers in various tissues of frogs (Rana ridibunda). Biomarkers selected for stress monitoring were malondialdehyde (MDA) and antioxidant defense system (ADS) constituent such as reduced glutathione (GSH)
Peter Eyer et al.
Biochemical pharmacology, 75(10), 2045-2053 (2008-04-04)
Isodimethoate is a thermal decomposition product that is present in usual pesticide formulations of dimethoate. Owing to its PO structure the compound is a direct anticholinesterase agent whose properties, to the best of our knowledge, are presented here for the
Yiou Pan et al.
Chemico-biological interactions, 188(3), 553-557 (2010-08-10)
The degree of insecticide resistance, acetylcholinesterase (AChE) activity kinetics, AChE gene expression and the cDNA sequence of AChE gene were investigated in resistant and relatively susceptible strains of the cotton aphids, Aphis gossypii (Glover). The resistant strain (ORR) exhibited 53.28-fold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service