Skip to Content
MilliporeSigma

Skip To

36640

Procymidone

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H11Cl2NO2
Molecular Weight:
284.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
251-233-1
Beilstein/REAXYS Number:
1539058
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC12CC1(C)C(=O)N(C2=O)c3cc(Cl)cc(Cl)c3

InChI

1S/C13H11Cl2NO2/c1-12-6-13(12,2)11(18)16(10(12)17)9-4-7(14)3-8(15)5-9/h3-5H,6H2,1-2H3

InChI key

QXJKBPAVAHBARF-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
683673235545684
description

PESTANAL®, analytical standard

description

PESTANAL®, analytical standard

description

VETRANAL®, analytical standard

description

PESTANAL®, analytical standard

format

neat

format

neat

format

neat

format

neat

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

PESTANAL®

product line

VETRANAL®

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

application(s)

agriculture

application(s)

forensics and toxicology
veterinary

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Procymidone


Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters