Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32498

Supelco

Silthiofam

PESTANAL®, analytical standard

Synonym(s):

N-Allyl-4,5-dimethyl-2-(trimethylsilyl)thiophene-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NOSSi
CAS Number:
Molecular Weight:
267.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1sc(c(c1C)C(=O)NCC=C)[Si](C)(C)C

InChI

1S/C13H21NOSSi/c1-7-8-14-12(15)11-9(2)10(3)16-13(11)17(4,5)6/h7H,1,8H2,2-6H3,(H,14,15)

InChI key

MXMXHPPIGKYTAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silthiofam is a thiophenecarboxamide fungicide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Silthiofam may be used as a reference standard in the determination of silthiofam in water samples using high-performance liquid chromatography with electrospray ionization-tandem mass spectrometry (HPLC-ESI-MS/MS).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia, 6(4), 357-366 (2014)
Process research on the synthesis of silthiofam: A novel fungicide for wheat
Phillips G, et al.
Organic Process Research & Development, 6(4), 357-366 (2002)
A simple and sensitive method for simultaneous analysis of multiresidual fungicides in water samples by HPLC-ESI-MS/MS
Wang Y, et al.
Analytical Methods : Advancing Methods and Applications, 4(12), 4146-4154 (2012)
Yin-Liang Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 989, 11-20 (2015-03-24)
A modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) method using multi-walled carbon nanotubes (MWCNTs) as a reversed-dispersive solid phase extraction (r-dSPE) material combined with ultra-high liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) was developed for the simultaneous determination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service