Skip to Content
MilliporeSigma
All Photos(1)

Documents

35597

Supelco

PCB No 15

analytical standard

Synonym(s):

4,4′-Dichlorobiphenyl, 4,4′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2
CAS Number:
Molecular Weight:
223.10
Beilstein:
2044700
Ballschmiter Number:
15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(cc1)-c2ccc(Cl)cc2

InChI

1S/C12H8Cl2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

YTBRNEUEFCNVHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Chu et al.
Water research, 36(9), 2187-2194 (2002-07-11)
Surfactant and organic solvents have individually been shown useful in assisting the solubilization of hydrophobic organics out of contaminated soil and promoting UV-induced photodecay at a succeeding treatment process. A newly proposed process is to use the mixtures of surfactant
Uptake of 4-chlorobiphenyl and 4,4'-dichlorobiphenyl in six species of plant tissue cultures.
K Yamamoto et al.
Bulletin of environmental contamination and toxicology, 28(6), 728-732 (1982-06-01)
R Mavoungou et al.
The Science of the total environment, 101(3), 263-268 (1991-01-15)
Anaerobic cultures containing 4,4'-dichlorobiphenyl (4,4'CB) were inoculated with various environmental samples. The degradation of the substrate was followed by gas chromatographic analysis. Three of the cultures tested showed an ability to dehalogenate the substrate, as judged by the presence of
L van Bree et al.
Toxicology, 42(2-3), 259-274 (1986-12-15)
Both 4,4'-dichlorobiphenyl (4,4'-DCB) and 3-methylcholanthrene (3-MC) caused a substantial increase of phenacetin-induced hepatic glutathione (GSH) depletion, whereas phenobarbital (PB) had no effect, suggesting that 4,4'-DCB possesses cytochrome P-448 inducing activity. The O-deethylation of phenacetin by liver microsomes from control and
C Edwin Garner et al.
Toxicology and applied pharmacology, 216(1), 157-167 (2006-06-21)
The fate of selected polychlorobiphenyls (PCBs) was investigated following single dermal administration (0.4 mg/kg) to determine the effects of chlorine content and position on the disposition of PCBs following dermal absorption. Single dermal doses of (14)C-labeled mono-, di-, tetra- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service