MilliporeSigma
  • Microbial dehalogenation of 4,4'-dichlorobiphenyl under anaerobic conditions.
Microbial dehalogenation of 4,4'-dichlorobiphenyl under anaerobic conditions.
The Science of the total environment (1991-01-15)
R Mavoungou, R Massé, M Sylvestre
ABSTRACT
Anaerobic cultures containing 4,4'-dichlorobiphenyl (4,4'CB) were inoculated with various environmental samples. The degradation of the substrate was followed by gas chromatographic analysis. Three of the cultures tested showed an ability to dehalogenate the substrate, as judged by the presence of 4-chlorobiphenyl and biphenyl in those media inoculated with them.
Product Description
Supelco
PCB No 15, analytical standard