Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35592

Supelco

PCB No 2

analytical standard

Synonym(s):

3-Chlorobiphenyl, 3-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9Cl
CAS Number:
Molecular Weight:
188.65
Beilstein:
1863135
Ballschmiter Number:
2
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1cccc(c1)-c2ccccc2

InChI

1S/C12H9Cl/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H

InChI key

NMWSKOLWZZWHPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls (PCBs) are groups of chemical compounds which are persistent in the environment and are considered highly toxic to humans and animals. PCB No 2 may be used for environmental analysis and precise quality control of food and feed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Fava et al.
Applied microbiology and biotechnology, 36(2), 240-245 (1991-11-01)
A mixed bacterial culture obtained from polychlorinated-biphenyl-contaminated river sediments proved capable of degrading 3-chlorobiphenyl (3-CB) under aerobic laboratory conditions. Almost total mineralization of 150 mg/l of 3-CB occurred when, after 3 days of incubation, the mineral medium was supplied with
B P Maliwal et al.
Journal of lipid research, 23(3), 474-479 (1982-03-01)
The uptake, distribution, and exchange of chlorinated hydrocarbon insecticides (dieldrin and chlordecone) and biphenyls (2,4,5-2',4',5'-hexachlorobiphenyl and 3-chlorobiphenyl) among human lipoproteins was examined by fluorescence quenching, gel filtration, and ultrafiltration. The chlorinated hydrocarbons were rapidly taken up from solution or silica
M W Kennedy et al.
Biochemical pharmacology, 29(5), 727-736 (1980-03-01)
In vitro rat hepatic microsomal metabolism of the monochlorobiphenyls (MCBs) 2-, 3- and 4-chlorobiphenyl, has been investigated as a model for the metabolism of polychlorinated biphenyl pollutants. MCB metabolism was catalyzed by cytochrome P-450, as indicated by a dependence on
A T Nielsen et al.
Environmental microbiology, 2(1), 59-68 (2001-03-13)
A flow cell-grown model consortium consisting of two organisms, Burkholderia sp. LB400 and Pseudomonas sp. B13(FR1), was studied. These bacteria have the potential to interact metabolically because Pseudomonas sp. B13(FR1) can metabolize chlorobenzoate produced by Burkholderia sp. LB400 when grown
R H Adams et al.
Applied and environmental microbiology, 58(2), 647-654 (1992-02-01)
Recombinant Pseudomonas sp. strain CB15, which grows on 3-chlorobiphenyl (3CB), was constructed from Pseudomonas sp. strain HF1, which grows on 3-chlorobenzoate, and from Acinetobacter sp. strain P6, which grows on biphenyl, by using a continuous amalgamated culture apparatus. DNA from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service