Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32849

Supelco

Amisulbrom

PESTANAL®, analytical standard

Synonym(s):

3-(3-Bromo-6-fluoro-2-methylindol-1-ylsulfonyl)-N,N-dimethyl-1H-1,2,4-triazole-1-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13BrFN5O4S2
CAS Number:
Molecular Weight:
466.31
Beilstein:
11328670
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)S(=O)(=O)n1cnc(n1)S(=O)(=O)n2c(C)c(Br)c3ccc(F)cc23

InChI

1S/C13H13BrFN5O4S2/c1-8-12(14)10-5-4-9(15)6-11(10)20(8)25(21,22)13-16-7-19(17-13)26(23,24)18(2)3/h4-7H,1-3H3

InChI key

BREATYVWRHIPIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amisulbrom is classified under the triazole group of fungicides. It contains a sulfone functional group with potent bioactivity.

Application

Amisulbrom may be used as an analytical reference standard for the determination of the analyte in fruits and vegetables by Dutch mini-Luke extraction and liquid chromatography-triple quadrupole mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of tobacco bacterial wilt with sulfone derivatives containing an 1, 3, 4-oxadiazole moiety.
Xu WM, et al.
Journal of Agricultural and Food Chemistry, 60(4), 1036-1041 (2012)
Miniaturisation and optimisation of the Dutch mini-Luke extraction method for implementation in the routine multi-residue analysis of pesticides in fruits and vegetables.
Lozano A, et al.
Food Chemistry, 192(13), 668-681 (2016)
Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography?Tandem Mass Spectrometry.
Shin Y, et al.
Journal of Agricultural and Food Chemistry, 66(13), 3550-3560 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service