Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32457

Supelco

Aminopyralid

PESTANAL®, analytical standard

Synonym(s):

4-Amino-3,6-dichloropicolinic acid, 4-Amino-3,6-dichloropyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2N2O2
CAS Number:
Molecular Weight:
207.01
Beilstein:
11403922
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1cc(Cl)nc(C(O)=O)c1Cl

InChI

1S/C6H4Cl2N2O2/c7-3-1-2(9)4(8)5(10-3)6(11)12/h1H,(H2,9,10)(H,11,12)

InChI key

NIXXQNOQHKNPEJ-UHFFFAOYSA-N

General description

Aminopyralid is a pyridine carboxylic acid auxinic herbicide, and is used pastures, non-crop and rangelands areas.
It has been used to control Canada Thistle (Cirsium arvense), a weed in a variety of crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canada thistle (Cirsium arvense) control with aminopyralid in range, pasture, and noncrop areas.
Enloe, Stephen F., et al.
Weed Technology, 21(4), 890-894 (2007)
Aminopyralid and clopyralid absorption and translocation in Canada thistle (Cirsium arvense).
Bukun, Bekir, et al.
Weed Sci., 57 (1), 10-15 (2009)
Patrick L Tomco et al.
Environmental toxicology and chemistry, 35(2), 485-493 (2015-08-28)
High-latitude regions experience unique conditions that affect the degradation rate of agrochemicals in the environment. In the present study, data collected from 2 field sites in Alaska, USA (Palmer and Delta) were used to generate a kinetic model for aminopyralid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service