Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37047

Supelco

Fluxapyroxad

PESTANAL®, analytical standard

Synonym(s):

3-(Difluoromethyl)-1-methyl-N-(3′,4′,5′-trifluorobiphenyl-2-yl)pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12F5N3O
CAS Number:
Molecular Weight:
381.30
Beilstein:
15702063
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cn1cc(C(=O)Nc2ccccc2-c3cc(F)c(F)c(F)c3)c(n1)C(F)F

InChI

1S/C18H12F5N3O/c1-26-8-11(16(25-26)17(22)23)18(27)24-14-5-3-2-4-10(14)9-6-12(19)15(21)13(20)7-9/h2-8,17H,1H3,(H,24,27)

InChI key

SXSGXWCSHSVPGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fluxapyroxad belongs to carboxamides group of chemicals. It is a succinate dehydrogenase inhibitor (SDHI) in complex II of the mitochondrial respiratory chain.

Application

Fluxapyroxad may be used as a reference standard for the determination of fluxapyroxad in foodstuffs by ultra-performance liquid chromatography coupled with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective monitoring of fluxapyroxad and its three biologically active metabolites in vegetables, fruits, and cereals by optimized QuEChERS treatment based on UPLC-MS/MS.
Chen X, et al.
Journal of Agricultural and Food Chemistry, 64(46), 8935-8943 (2016)
Quantifying the non-fungicidal effects of foliar applications of fluxapyroxad (Xemium) on stomatal conductance, water use efficiency and yield in winter wheat.
Smith J, et al.
Communications in agricultural and applied biological sciences, 78(3), 523-535 (2013)
Sensitivity of Rhizoctonia solani causing rice sheath blight to fluxapyroxad in China.
Chen Y, et al.
European Journal of Plant Pathology, 140(3), 419-428 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service