Skip to Content
MilliporeSigma
Bisoctrizole Pharmaceutical Secondary Standard; Certified Reference Material

Bisoctrizole

Bisoctrizole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol]
Empirical Formula (Hill Notation):
C41H50N6O2
CAS Number:
Molecular Weight:
658.87
EC Number:

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique