Skip to Content
MilliporeSigma
All Photos(2)

Documents

27097

Supelco

Chromoionophore VII

Selectophore

Synonym(s):

9-Dimethylamino-5-[4-(15-butyl-1,13-dioxo-2,14-dioxanonadecyl)phenylimino]benzo[a]phenoxazine, ETH 5418

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H57N3O5
CAS Number:
Molecular Weight:
719.95
MDL number:
UNSPSC Code:
26111700

product line

Selectophore

Quality Level

General description

Chromoionophore VII (ETH 5418) is a lipophilic ionophore.
Visit our Sensor Applications portal to learn more.

Application

ETH 5418 was used for the designing of fluorescent microspheres.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

H+-selective chromoionophore used in a cation exchange system for the detn. of lead with lead ionophore

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel potentiometric and optical silver ion-selective sensors with subnanomolar detection limits.
Szigeti Z
Analytica Chimica Acta, 572 (1), 1-10 (2006)
Mass-produced ionophore-based fluorescent microspheres for trace level determination of lead ions.
Telting-Diaz
Analytical Chemistry, 74 (20), 5251-5256 (2002)
M. Lerchi et al.
Analytical Chemistry, 64, 1534-1534 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service