Skip to Content
MilliporeSigma
All Photos(2)

Documents

27087

Supelco

Chromoionophore II

Selectophore

Synonym(s):

9-Dimethylamino-5-[4-(16-butyl-2,14-dioxo-3,15-dioxaeicosyl)phenylimino]benzo[a]phenoxazine, ETH 2439

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H59N3O5
CAS Number:
Molecular Weight:
733.98
Beilstein:
6626911
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

SMILES string

CCCCC(CCCC)OC(=O)CCCCCCCCCCOC(=O)Cc1ccc(cc1)\N=C2\C=C3Oc4cc(ccc4N=C3c5ccccc25)N(C)C

InChI

1S/C46H59N3O5/c1-5-7-19-37(20-8-6-2)53-44(50)23-15-13-11-9-10-12-14-18-30-52-45(51)31-34-24-26-35(27-25-34)47-41-33-43-46(39-22-17-16-21-38(39)41)48-40-29-28-36(49(3)4)32-42(40)54-43/h16-17,21-22,24-29,32-33,37H,5-15,18-20,23,30-31H2,1-4H3/b47-41-

InChI key

SWOXJPWUWJDGGP-LOKWYOQLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromoionophore II (ETH 2439, 9-Dimethylamino-5-[4-(16-butyl-2,14-dioxo-3,15-dioxaeicosyl)phenylimino]benzo[a]phenoxazine) is a phenoxazine derivative.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Assay of Na+ activity with solvent polymeric optode membrane prepared with ETH 2439 and sodium ionophore V; Assay of Ca2+ activity with solvent polymeric optode membranes prepared from ETH 2439 and calcium ionophore I

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The potentiometric pH response of chromoionophores used in optical sensors.
LInder E, et.la.
Journal of Electroanalytical Chemistry, 352 (1), 309-312 (1993)
Hannah R Schwartz et al.
Cell reports, 31(12), 107800-107800 (2020-06-25)
When evaluating anti-cancer drugs, two different measurements are used: relative viability, which scores an amalgam of proliferative arrest and cell death, and fractional viability, which specifically scores the degree of cell killing. We quantify relationships between drug-induced growth inhibition and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service