Skip to Content
MilliporeSigma
All Photos(2)

Documents

27088

Supelco

Chromoionophore III

Selectophore

Synonym(s):

9-(Diethylamino)-5-[(2-octyldecyl)imino]benzo[a]phenoxazine, ETH 5350

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H55N3O
CAS Number:
Molecular Weight:
569.86
Beilstein:
5462368
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

SMILES string

CCCCCCCCC(CCCCCCCC)C\N=C1/C=C2Oc3cc(ccc3N=C2c4ccccc14)N(CC)CC

InChI

1S/C38H55N3O/c1-5-9-11-13-15-17-21-30(22-18-16-14-12-10-6-2)29-39-35-28-37-38(33-24-20-19-23-32(33)35)40-34-26-25-31(27-36(34)42-37)41(7-3)8-4/h19-20,23-28,30H,5-18,21-22,29H2,1-4H3/b39-35+

InChI key

ICVAQSQRYBGWQH-IGIMMJHKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromoionophore III (ETH 5350) is a pH sensitive dye, and is ratiometric that is the fluorescence spectrum has two peaks, and the ratio between these peaks changes according to the changing pH levels.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
ETH 5350 was used for the fabrication of optode membranes, which are sodium selective, used in order to optically determine sodium in pH-buffered human blood plasma.
Assay of Na+ activity with solvent polymeric optode membrane prepared from ETH 5350 and sodium ionophore V; Assay of ammonia with solvent polymeric optode membranes based on ETH 5350 and valinomycin (potassium ionophore I)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic microdrill as a modulated fluorescent pH sensor.
McNaughton
Journal of magnetism and magnetic materials, 293 (1), 696-701 (2005)
Characterization of sodium-selective optode membranes based on neutral ionophores and assay of sodium in plasma.
Seiler K
Clinical Chemistry, 37 (8), 1350-1355 (1991)
A A Galyean et al.
The Analyst, 143(2), 458-465 (2017-12-12)
Nanosensors present a biological monitoring method that is biocompatible, reversible, and nano-scale, and they offer many advantages over traditional organic indicators. Typical ionophore-based nanosensors incorporate nile-blue derivative pH indicators but suffer from photobleaching while quantum dot alternatives pose a potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service