Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

24539

Hamilton® syringe, 700 series, removable needle

750RN, volume 0.5 mL, needle size 22 ga (bevel tip), needle L 51 mm (2 in.)

Synonym(s):

Hamilton® Syringe 750RN, 500UL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500

material

stainless steel needle (point style 2)

needle size

22 ga (bevel tip)

feature

needle type RN (removable needle)
point style 2

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80830

needle L

51 mm (2 in.)

volume

0.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.

Application

It has been used during greenhouse testing done for evaluating transformed lines expressing FsTR1101 gene for resistance to the spread of Fusarium. It was also used in determining the bubble stability and for optical imaging of bubbles of Class II Hydrophobins from Trichoderma reesei.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface properties of class II hydrophobins from Trichoderma reesei and influence on bubble stability.
Cox, Andrew R., et al.
Langmuir, 23.15, 7995-8002 (2007)
Engineering deoxynivalenol metabolism in wheat through the expression of a fungal trichothecene acetyltransferase gene.
Okubara, Patricia, et al.
Tag. Theoretical and Applied Genetics. Theoretische Und Angewandte Genetik, 106.1, 74-83 (2002)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service