Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16101

Sigma-Aldrich

Resorcinol

meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39023402
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 21.1 °C)

Assay

98.5-100.5% (calc. to the dried substance)

form

solid

autoignition temp.

1126 °F

quality

meets analytical specification of Ph. Eur., BP

impurities

acidity or alkalinity, complies
related subst., complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.01% free acid (as H2SO4)
≤0.01% free alkali (as NH3)
≤0.01% pyrocatechol

ign. residue

≤0.05% (as SO4)

loss

≤1.0% loss on drying (on silica gel)

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

solubility

water: soluble

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

suitability

complies for appearance of solution
complies for identity

application(s)

pharmaceutical (small molecule)

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Resorcinol has been used in resorcinol staining for visualizing the gangliosides. It has also been used for estimating sialic acids by colorimetric assay.

Biochem/physiol Actions

Resorcinol is an aromatic alcohol serves as an antiseptic. It reduces pain from painful nodules in patients suffering from Hidradenitis suppurativa (HS).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Masson et al.
The Biochemical journal, 388(Pt 2), 537-544 (2005-01-19)
Alterations in proliferation and hypertrophy of renal mesangial cells are typical features of diabetic nephropathy. The HP (hexosamine pathway) has been proposed as a biochemical hypothesis to explain microvascular alterations due to diabetic nephropathy; however, involvement of HP in the
J Boer et al.
Clinical and experimental dermatology, 35(1), 36-40 (2009-06-25)
Hidradenitis suppurativa (HS) is a chronic, inflammatory skin disease characterized by abscess formation localized to apocrine sweat gland-bearing skin. The most important factor in patients' overall assessment of disease severity is pain. The duration of abscesses takes days to weeks
Ariana Kupai et al.
Frontiers in cell and developmental biology, 8, 241-241 (2020-04-25)
Lysine methylation facilitates protein-protein interactions through the activity of methyllysine (Kme) "reader" proteins. Functions of Kme readers have historically been studied in the context of histone interactions, where readers aid in chromatin-templated processes such as transcription, DNA replication and repair.
Anton V Gulevich et al.
Journal of the American Chemical Society, 134(12), 5528-5531 (2012-03-15)
The efficient Pd-catalyzed double-fold C-H oxygenation of arenes into resorcinols using the newly developed 2-pyrimidyldiisopropylsilyl (PyrDipSi) directing group is described. Its use allows for the sequential introduction of OAc and OPiv groups in a one-pot manner to produce orthogonally protected
Maurizio Scaltriti et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(17), 4508-4513 (2012-06-22)
Many kinases and hormone receptors, important for cancer cell proliferation and survival, bind to and are dependent on the Hsp90 cycle for their folding and maturation. This provides the rationale for the development of small-molecule ATP competitors that, inhibiting Hsp90

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service