Skip to Content
MilliporeSigma
All Photos(2)

Documents

331635

Sigma-Aldrich

Tetramethylammonium hydroxide solution

25 wt. % in H2O

Synonym(s):

TMAH solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OH)
CAS Number:
Molecular Weight:
91.15
Beilstein:
3558708
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

17.5 mmHg ( 20 °C)

Quality Level

form

liquid

concentration

25 wt. % in H2O

refractive index

n20/D 1.3806

density

1.016 g/mL at 25 °C

SMILES string

[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.H2O/c1-5(2,3)4;/h1-4H3;1H2/q+1;/p-1

InChI key

WGTYBPLFGIVFAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylammonium hydroxide solution is commonly used as a strong base and nucleophilic source in organic synthesis reactions.

Application

Tetramethylammonium hydroxide is a quaternary ammonium salt generally used as an anisotropic etchant for silicon due to its high silicon etching rate.
Tetramethylammonium hydroxide solution (25 wt.% solution in water) may be used as a base for pH adjustment to obtain hexagonal mesoporous aluminophosphate (TAP).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1 Dermal - STOT SE 1

Target Organs

Central nervous system, Liver,thymus

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium-mediated carboxylation of aryl halides (triflates) or benzyl halides using [13 C]/[11 C] carbon monoxide with tetrabutylammonium hydroxide or trimethylphenylammonium hydroxide
Karimi F, et al.
Journal of the Chemical Society. Perkin Transactions 1, 20, 2256-2259 (2002)
Synthesis of mesoporous hexagonal titanium aluminophosphate molecular sieves and their catalytic applications
Kapoor MP & Raj A.
Applied Catalysis A: General, 203(2), 311-319 (2000)
Tetramethylammonium Hydroxide
Knight J
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Study on anisotropic silicon etching characteristics in various surfactant-added tetramethyl ammonium hydroxide water solutions.
Yang CR, et al.
Journal of Micromechanics and Microengineering, 15(11), 2028-2028 (2005)
The characteristic behavior of TMAH water solution for anisotropic etching on both silicon substrate and SiO 2 layer.
Chen PH, et al.
Sensors and actuators A, Physical, 93(2), 132-137 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service