Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.07590

SAFC

Resorcinol

extra pure

Synonym(s):

Resorcinol, 1,3-Dihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)2
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-585-2

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

605 °C

expl. lim.

1.4 % (v/v)

pH

4-6 (20 °C, 100 g/L in H2O)

bp

281 °C/1013 hPa

mp

109-111 °C

transition temp

flash point 127 °C

solubility

1000 g/L

density

1.28 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

15-25°C

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Resorcinol extra pure is suitable for use in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service