Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13198

Supelco

Redox standard

250 mV

Synonym(s):

250 mV 500ml Redox standard, Redox standard 250 mV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

description

redox potential: 250 mV ±5 mV, 25°C

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Redox standard has a potential of 250 mV ±5 mV. It refers to the potential of the platinum electrode versus the Ag/AgCl reference electrode in 3M potassium chloride.

Application

Redox standard acts as a control standard rather than a calibration standard which controls the functionality of the sensing and reference electrodes.

Features and Benefits

  • Broad range of values
  • Very high specifications (±5mV)
  • Detailed safety data sheet (SDS) available
  • Certified with proven verifiable accuracy and uncertainty of measurement
  • Guaranteed stability throughout its entire shelf life

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service