Skip to Content
MilliporeSigma

06949

Conductivity Standard

conductivity 1000 μS/cm at 25 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

≤2.5% KCl (T)

conductivity

990.0-1010.0 μS/cm at 25 °C

density

0.995-1.014 g/cm3 at 20 °C

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

format

single component solution

Application

Analytical standard for conductivity meter calibration.


Still not finding the right product?

Explore all of our products under Conductivity Standard


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2 - STOT RE 2 Oral

target_organs

Liver,Kidney

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library