Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

114553

Sigma-Aldrich

Thymolphthalein

ACS reagent, Dye content 95 %

Synonym(s):

5′,5′′-Diisopropyl-2′,2′′-dimethylphenolphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30O4
CAS Number:
Molecular Weight:
430.54
Beilstein:
359413
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

solid

composition

Dye content, 95%

clarity of soln

passes test

technique(s)

titration: suitable

visual transition interval

8.8-10.5, colorless to blue

mp

251-253 °C (lit.)

λmax

396 nm (2nd)
592 nm

SMILES string

CC(C)c1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

InChI

1S/C28H30O4/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(31)32-28)24-14-21(16(3)4)26(30)12-18(24)6/h7-16,29-30H,1-6H3

InChI key

LDKDGDIWEUUXSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thymolphthalein (5′, 5′′-Diisopropyl-2′, 2′′-dimethylphenolphthalein) is an acid-base indicator which is colorless in solutions with a pH below 8.5 and blue in solutions with a pH above 9.5. Thymolphthalein has been used as an indicator in periodate-thiosulfate reaction.

Application

Thymolphthalein has been used as a pH indicator in [14C] urea breath test to determine when the CO2 trap is full.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of exercise on VLDL-triglyceride oxidation and turnover.
Sondergaard E
American Journal of Physiology. Endocrinology and Metabolism, 300(5), E939-E944 (2011)
Rhesus monkey (Macaca mulatta) model of Helicobacter pylori: noninvasive detection and derivation of specific-pathogen-free monkeys.
Laboratory Animal Science, 49(2), 197-201 (1999)
Basal and insulin mediated VLDL-triglyceride kinetics in type 2 diabetic men.
S?rensen LP
Diabetes, 60(1), 88-96 (2011)
American Chemical Society, American Chemical Society. Committee on Analytical Reagents
Reagent Chemicals: Specifications and Procedures (2006)
Bassam Z. Shakhashiri
Chemical Demonstrations: A Handbook for Teachers of Chemistry, Volume 3, 3 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service