Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
powder
Quality Segment
technique(s)
microbe id | staining: suitable
loss
≤5% loss on drying
pH
3.87 (30 °C, 7.14 g/L)
mp
>340 °C
solubility
methanol: 0.01 g/10 mL, blue to very deep blue
density
0.71 g/cm3 at 29 °C
εmax
≥350 at 603-613 nm
suitability
suitable for microscopy
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Na+].[Na+].[O-]S(=O)(=O)c1ccc2NC(\C(=O)c2c1)=C3\Nc4ccc(cc4C3=O)S([O-])(=O)=O
InChI
1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;
InChI key
KHLVKKOJDHCJMG-QDBORUFSSA-L
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| description suitable for microscopy | description analytical standard | description United States Pharmacopeia (USP) Reference Standard | description certified by the BSC, Dye content 85 % |
| technique(s) microbe id | staining: suitable | technique(s) gas chromatography (GC): suitable, HPLC: suitable | technique(s) - | technique(s) - |
| suitability suitable for microscopy | suitability - | suitability - | suitability - |
| form powder | form - | form - | form solid |
| application(s) diagnostic assay manufacturing | application(s) cleaning products | application(s) pharmaceutical (small molecule) | application(s) diagnostic assay manufacturing |
| solubility methanol: 0.01 g/10 mL, blue to very deep blue | solubility - | solubility - | solubility - |
| storage temp. room temp | storage temp. - | storage temp. - | storage temp. room temp |
Still not finding the right product?
Explore all of our products under Indigo carmine
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

