Skip to Content
MilliporeSigma

89360

Thymolphthalein

ACS reagent

Synonym(s):

5′,5′′-Diisopropyl-2′,2′′-dimethylphenolphthalein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H30O4
CAS Number:
Molecular Weight:
430.54
UNSPSC Code:
12352200
NACRES:
NA.21
PubChem Substance ID:
EC Number:
204-729-7
Beilstein/REAXYS Number:
359413
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

form

solid

technique(s)

UV/Vis spectroscopy: suitable, titration: suitable

ign. residue

≤1%

mp

248-254 °C, 251-253 °C (lit.)

SMILES string

CC(C)c1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

InChI

1S/C28H30O4/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(31)32-28)24-14-21(16(3)4)26(30)12-18(24)6/h7-16,29-30H,1-6H3

InChI key

LDKDGDIWEUUXSH-UHFFFAOYSA-N

General description

Thymolphthalien is an acid-base indicator which is colourless in acidic form and deep blue in basic form. Acidic form retains hydrogen on each hydroxyl group. It is suitable in preparing disappearing ink.

Application

Thymolphthalein was suitable as indicator during titration of hyamine hydroxide.


Still not finding the right product?

Explore all of our products under Thymolphthalein


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library