Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

114537

Sigma-Aldrich

Phenol Red sodium salt

ACS reagent, Dye content 90 %

Synonym(s):

Phenolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13NaO5S
CAS Number:
Molecular Weight:
376.36
Beilstein:
6260026
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder or crystals

composition

Dye content, 90%

clarity of soln

passes test

technique(s)

titration: suitable

color

red to very dark red

visual transition interval

6.8-8.2, yellow to red
6.8-8.2, yellow to red

mp

285 °C (dec.) (lit.)

λmax

423 nm

ε (extinction coefficient)

≥19000 at 422-426 nm in methanol
≥900 at 256-269 nm in methanol

storage temp.

room temp

SMILES string

[Na+].Oc1ccc(cc1)C2(OS(=O)(=O)c3ccccc23)c4ccc([O-])cc4

InChI

1S/C19H14O5S.Na/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19;/h1-12,20-21H;/q;+1/p-1

InChI key

HKHYOKBQJILTEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phenol Red has been used for preparation of phosphate buffer saline (PBS)-with phenol red for microfluidic assay for simultaneous culture of multiple cell types. It has also been used for horseradish peroxidase-phenol red (HRP-PR) H2O2 (hydrogen peroxide) detection assay to identify redox cycling compounds.
Suitable for use as a pH indicator.

Biochem/physiol Actions

Phenol Red is a pH indicator which changes color from yellow to red.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling the NIH Small Molecule Repository for compounds that generate H2O2 by redox cycling in reducing environments.
Soares KM, et al.
Assay and Drug Development Technologies, 8, 152-174 (2010)
Microfluidic assay for simultaneous culture of multiple cell types on surfaces or within hydrogels.
Shin Y, et al.
Nature Protocols, 7, 1247-1259 (2012)
TiO2 nanotube arrays of 1000 ?m length by anodization of titanium foil: phenol red diffusion.
Paulose M, et al.
The Journal of Physical Chemistry C, 111, 14992-14997 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service