Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07241

Supelco

Phenol red

pH indicator, ACS

Synonym(s):

Phenol red

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4OH)2C7H4SO3
CAS Number:
Molecular Weight:
354.38
MDL number:
UNSPSC Code:
12171502
EC Index Number:
205-609-7
NACRES:
NA.21

Product Name

Phenol red, indicator ACS

grade

ACS reagent

Quality Level

form

solid

loss

≤1% loss on drying, 110°C

visual transition interval

1.2-3.0, brown-orange to yellow(1st transition range)
6.5-8.0, brownish-yellow to red-violet(2nd transition range)

mp

>300 °C

bulk density

200‑300 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3ccc(cc3)O)c2ccc(cc2)O

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

General description

Phenol red (phenolsulfonphthalein, PH R) belongs to the class of triphenylmethane dyes. It is a waterͰsoluble anionic dye, mostly used as a pH indicator in cell biology laboratories. Also, in different protocols, it is used to detect H2O2(Hydrogen peroxide).

Application

Phenol red can be used as a:
  • Dye in the preparation of phenol red/n-Si photodiode [phenol red/n-type silicon wafer] by spin coating technique. The device is appropriate for photo-detection applications.
  • Color-changing dye in the design of fiber optic pH sensor because as the acidity increases, the dye′s absorption properties shift from the green to the blue section of the spectrum.
  • Indicator in the determination of freshwater pH.

Features and Benefits

Phenol red is used to detect pH changes in cell culture,
  • pH is 6.4 or below yellow color
  • pH is 8.2 or above red color.

Analysis Note

Appearance of solution: passes test
1st transition range: pH 1.2 - pH 3.0brownish orange - yellow
2nd transition range: pH 6.5 - pH 8.0brownish yellow - red violet
Loss on drying (110 °C): ≤ 1
Transition range (according to ACS): passes test
ACS conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service